Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPS20 Rabbit anti-Human, Clone: SC65-08, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP267628
Description
RPS20 Monoclonal specifically detects RPS20 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
RPS20 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
40S ribosomal protein S20, FLJ27451, MGC102930, ribosomal protein S20 | |
Rabbit | |
Protein A purified | |
DNA replication Transcription Translation and Splicing | |
6224 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
SC65-08 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
RPS20 | |
Synthetic peptide within Human RPS20 aa 36-80/ 119. (SwissProt: P60866 Human; SwissProt: P60867 Mouse; SwissProt: P60868 Rat) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction