Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPS21 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551914
Description
Immunogen sequence: MQNDAGEFVD LYVPRKCSAS NRIIGAKDHA SIQMNVAEVD KVTGRFNGQF KTYAICGAIR RMGESDDSIL RLAKADGIVS K Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
Differentiated embryo-chondrocyte expressed gene 1 (DEC1) is an important protein involved in embryonic cell differentiation, proliferation, and apoptosis. It has also recently been shown to be regulated by hypoxia. The loss of DEC1 expression may be an early event in the development of lung cancer, while DEC1 may be up-regulated by hypoxia in other cancers and in more extreme hypoxia it may have a role in cell death.
Specifications
| RPS21 | |
| Polyclonal | |
| Unconjugated | |
| RPS21 | |
| 1810049N11Rik; 2410030A14Rik; 40S ribosomal protein S21; 8.2 kDa differentiation factor; HLDF; human leukemia differentiation factor; LOC615178 protein; ribosomal protein S21; rps21; S21; Small ribosomal subunit protein eS21; zgc:56642; zgc:86816 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6227, 66481, 81775 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P05765, P63220, Q9CQR2 | |
| RPS21 | |
| Recombinant protein corresponding to Human RPS21. Recombinant protein control fragment (Product #RP-102214). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction