Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPS21 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103534
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103534 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103534 Supplier Thermo Scientific Supplier No. PA5103534
Add to Cart
Add to Cart

RPS21 Polyclonal Antibody

RPS21 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen RPS21
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RPS21
Gene Accession No. P05765, P63220, Q9CQR2
Gene Alias 1810049N11Rik; 2410030A14Rik; 40S ribosomal protein S21; 8.2 kDa differentiation factor; HLDF; human leukemia differentiation factor; LOC615178 protein; ribosomal protein S21; rps21; S21; Small ribosomal subunit protein eS21; zgc:56642; zgc:86816
Gene Symbols RPS21
Host Species Rabbit
Immunogen A synthesized peptide derived from human RPS21(Accession P63220), corresponding to amino acid residues V39-F83.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6227, 66481, 81775
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.