Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPS24 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588361
Description
Immunogen sequence: MNDTVTIRTR KFMTNRLLQR KQMVIDVLHP GKATVPKTEI REKLAKMYKT TPDVIFVFGF RTHFGGGKTT GFGMIYDSLD YAKKNEPKHR LARHGLYEKK KTSRKQRKER KNRMKKVRGT AKANVGAGKK; Positive Samples: U-87MG, 293T, HeLa, Jurkat, Mouse liver, Mouse pancreas, Rat liver.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. RPS24 is a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S24E family of ribosomal proteins. It is located in the cytoplasm. Multiple transcript variants encoding different isoforms have been found for this gene. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
RPS24 | |
Polyclonal | |
Unconjugated | |
RPS24 | |
40S ribosomal protein S24; DBA3; DKFZp686N1586; MRP S24; ribosomal protein S24; ribosomal protein S24, isoform 1; RPS24; Rps24 protein; S24; Small ribosomal subunit protein eS24; wu:fa66b02; zgc:103596; zgc:109740 | |
Rabbit | |
Affinity chromatography | |
RUO | |
20088, 6229, 81776 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P62847, P62849, P62850 | |
RPS24 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-130 of human RPS24 (NP_1489821). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction