Learn More
Invitrogen™ RPS25 Polyclonal Antibody

Description
Positive test controls include: A431, BxPC-3, A549, HeLa, K-562, Mouse liver, Mouse pancreas, Rat liver. Immunogen sequence: FDKATYDKLC KEVPNYKLIT PAVVSERLKI RGSLARAALQ ELLSKGLIKL VSKHRAQVIY TRNTKGGDAP AAGEDA.
Specifications
Specifications
| Antigen | RPS25 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.6 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | RPS25 |
| Gene Accession No. | P62851, P62852, P62853 |
| Gene Alias | 2810009D21Rik; 40S ribosomal protein S25; ribosomal protein s25; RPS25; S25; Small ribosomal subunit protein eS25 |
| Gene Symbols | RPS25 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.