Learn More
Invitrogen™ RPS26 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585689
Description
Keep as concentrated solution. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (100%), Xenopus laevis (100%), Pig (100%), Sheep (100%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (100%). Positive Control: MCF-7, MDA-MB-231. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S26E family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
RPS26 | |
Polyclonal | |
Unconjugated | |
RPS26 | |
40S ribosomal protein S26; DBA10; ribosomal protein S26; Rps26; S26; Small ribosomal subunit protein eS26 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27139, 27370, 6231 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P62854, P62855, P62856 | |
RPS26 | |
Recombinant protein encompassing a sequence within the N-terminus region of human RPS26. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.