Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPS26 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590218
Description
Immunogen sequence: MTKKRRNNGR AKKGRGHVQP IRCTNCARCV PKDKAIKKFV IRNIVEAAAV RDISEASVFD AYVLPKLYVK LHYCVSCAIH SKVVRNRSRE ARKDRTPPPR; Positive Samples: U-251MG, LO2, Raji, DU145, mouse brain, mouse heart, mouse kidney, mouse liver.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S26E family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
RPS26 | |
Polyclonal | |
Unconjugated | |
RPS26 | |
40S ribosomal protein S26; DBA10; ribosomal protein S26; Rps26; S26; Small ribosomal subunit protein eS26 | |
Rabbit | |
Affinity chromatography | |
RUO | |
27139, 27370, 6231 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.96 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P62854, P62855, P62856 | |
RPS26 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human RPS26 (NP_0010202). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction