Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPS26 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | RPS26 |
---|---|
Concentration | 0.15 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-9826
![]() ![]() |
Proteintech Group Inc
149091AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S26E family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.Specifications
RPS26 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P62854, P62855, P62856 | |
27139, 27370, 6231 | |
RPS26 Fusion Protein Ag6706 | |
Primary | |
-20°C | |
RPS26 |
0.15 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
40S ribosomal protein S26, ribosomal protein S26, RPS26 | |
Rps26 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title