Learn More
Invitrogen™ RPS28 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Supplier:  Invitrogen™ PA545721
Catalog No. PIPA545721
Only null left
Add to Cart
Description
Description
Peptide sequence: RTGSQGQCTQ VRVEFMDDTS RSIIRNVKGP VREGDVLTLL ESEREARRLR Sequence homology: Cow: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 93%; Zebrafish: 100%.
TRPCs, mammalian homologs of the Drosophila transient receptor potential (trp) protein, are ion channels that are thought to mediate capacitative calcium entry into the cell. TRP-PLIK is a protein that is both an ion channel and a kinase. As a channel, it conducts calcium and monovalent cations to depolarize cells and increase intracellular calcium. As a kinase, it is capable of phosphorylating itself and other substrates. The kinase activity is necessary for channel function, as shown by its dependence on intracellular ATP and by the kinase mutants.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RPS28
Polyclonal
Unconjugated
RPS28
40S ribosomal protein S28; DBA15; fb07a10; ribosomal protein S28; rps28; S28; Small ribosomal subunit protein eS28; wu:fb07a10; wu:fb94f01; zgc:73367
Rabbit
Affinity Chromatography
RUO
6234
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry (Paraffin), Western Blot, Surface Plasmon Resonance
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
P62857
RPS28
Synthetic peptide directed towards the N-terminal of human RPS28 (aa 20-69).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.