Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPS29 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552051
Description
Immunogen sequence: MGHQQLYWSH PRKFGQGSRS CRVCSNRHGL IRKYGLNMCR QCFRQYAKDI GFIKL Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. RPS29 is a ribosomal protein that is a component of the 40S subunit and a member of the S14P family of ribosomal proteins. The protein, which contains a C2-C2 zinc finger-like domain that can bind to zinc, can enhance the tumor suppressor activity of Ras-related protein 1A (KREV1). It is located in the cytoplasm.
Specifications
| RPS29 | |
| Polyclonal | |
| Unconjugated | |
| RPS29 | |
| 40S ribosomal protein S29; DBA13; ribosomal protein S29; RPS29; S29; S29 ribosomal protein; Small ribosomal subunit protein uS14; zgc:113935 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6235 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P62273 | |
| RPS29 | |
| Recombinant protein corresponding to Human RPS29. Recombinant protein control fragment (Product #RP-89114). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction