Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPS3 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298931APC
Description
RPS3 Polyclonal antibody specifically detects RPS3 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
RPS3 | |
Polyclonal | |
Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation | |
40S ribosomal protein S3, FLJ26283, FLJ27450, IMR-90 ribosomal protein S3, MGC87870, ribosomal protein S3 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human RPS3. | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
6188 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunocytochemistry, Immunoprecipitation | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction