Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPS3 Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298930FR
Description
RPS3 Polyclonal specifically detects RPS3 in Human samples. It is validated for Western Blot.Specifications
RPS3 | |
Polyclonal | |
Western Blot | |
40S ribosomal protein S3, FLJ26283, FLJ27450, IMR-90 ribosomal protein S3, MGC87870, ribosomal protein S3 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human RPS3. | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
6188 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction