Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPS3 Antibody (RP159-1) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280043
Description
RPS3 Monoclonal specifically detects RPS3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
RPS3 | |
Monoclonal | |
1 mg/mL | |
Western Blot, ELISA, Immunohistochemistry | |
40S ribosomal protein S3, FLJ26283, FLJ27450, IMR-90 ribosomal protein S3, MGC87870, ribosomal protein S3 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Recognizes human RPS3. Detects a band of ∼32kDa by Western blot. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry | |
RP159-1 | |
Unconjugated | |
0.2 um-filtered solution in PBS, pH 7.4 | |
RPS3 | |
Recombinant human RPS3. | |
100 μg | |
Core ESC Like Genes, Stem Cell Markers | |
6188 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction