Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPS3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA586732
Description
Purity is > 95% by SDS-PAGE.
WD repeat domain 5 (WDR5) is a member of the WD repeat protein family, which is involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. WDR5, also known as BIG-3, is expressed in the developing growth plate, accelerates chondrocyte and osteoblast differentiation in vitro, and regulates osteoblast differentiation during embryonic bone development. WDR5 interacts with the pluripotency factor Oct4/POU5F1 and is required for the efficient formation of induced pluripotent stem (iPS) cells.
Specifications
RPS3 | |
Polyclonal | |
Unconjugated | |
RPS3 | |
40S ribosomal protein S3; D7Ertd795e; FLJ26283; FLJ27450; IMR-90 ribosomal protein S3; MGC87870; OK/SW-cl.26; OTTHUMP00000229804; OTTHUMP00000229805; OTTHUMP00000229874; OTTHUMP00000229877; OTTHUMP00000229878; OTTHUMP00000229879; OTTHUMP00000229880; OTTHUMP00000229882; OTTHUMP00000229883; OTTHUMP00000229886; ribosomal protein S3; Rps3; Rs_3; S3; Small ribosomal subunit protein uS3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
140654, 27050, 6188 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot, Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 0.05% sodium azide; pH 7.2 | |
P23396, P62908, P62909 | |
RPS3 | |
Synthetic peptide, corresponding to amino acids 198-243 of Human RPS3. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction