Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPS3 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA596153
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596153 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596153 Supplier Thermo Scientific Supplier No. PA596153

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

Immunogen sequence: MAVQISKKRK FVADGIFKAE LNEFLTRELA EDGYSGVEVR VTPTRTEIII LATRTQNVLG EKGRRIRELT AVVQKRFGFP EGSVELYAEK VATRGLCAIA QAESLRYKLL GGLAVRRACY GVLRFIMESG AKGCEVVVSG KLRGQRAKSM KFVDGLMIHS GDPVNYYVDT AVRHVLLRQG VLGIKVKIML PWDPTGKIGP KKPLPDHVSI VEPKDEILPT TPISEQKGGK PEPPAMPQPV PTA; Positive Samples: CEM, MCF-7, SW620, HT-29, Mouse liver, Mouse brain, Mouse heart; Cellular Location: Cytoplasm, Mitochondrion inner membrane, Nucleus, Peripheral membrane protein, cytoskeleton, nucleolus, spindle

WD repeat domain 5 (WDR5) is a member of the WD repeat protein family, which is involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. WDR5, also known as BIG-3, is expressed in the developing growth plate, accelerates chondrocyte and osteoblast differentiation in vitro, and regulates osteoblast differentiation during embryonic bone development. WDR5 interacts with the pluripotency factor Oct4/POU5F1 and is required for the efficient formation of induced pluripotent stem (iPS) cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPS3
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide, pH 7.3
Gene RPS3
Gene Accession No. P23396, P62908, P62909
Gene Alias 40S ribosomal protein S3, D7Ertd795e, FLJ26283, FLJ27450, IMR-90 ribosomal protein S3, MGC87870, OK/SW-cl.26, OTTHUMP00000229804, OTTHUMP00000229805, OTTHUMP00000229874, OTTHUMP00000229877, OTTHUMP00000229878, OTTHUMP00000229879, OTTHUMP00000229880, OTTHUMP00000229882, OTTHUMP00000229883, OTTHUMP00000229886, ribosomal protein S3, Rps3, Rs_3, S3, Small ribosomal subunit protein uS3
Gene Symbols RPS3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-243 of human RPS3 (NP_0009962)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140654, 27050, 6188
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.