Learn More
Invitrogen™ RPS3A Polyclonal Antibody

Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji, Neuro 2A, GL261, C8D30, NIH-3T3, BCL-1, Raw264.7, C2C12, PC-12, Rat2. Predicted reactivity: Rat (98%), Zebrafish (92%), Japanese Medaka (93%), Xenopus laevis (96%), Pig (97%), Chicken (96%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | RPS3A |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | RPS3A |
| Gene Accession No. | P49242, P61247, P97351 |
| Gene Alias | 40S ribosomal protein S3a; 40S ribosomal protein S3a {ECO:0000255; 40S ribosomal protein S3b; FTE1; Fte-1; HAMAP-Rule:MF_03122}; MFTL; Protein TU-11; ribosomal protein S3a; ribosomal protein S3A1; RPS3A; Rps3a1; S3A; Small ribosomal subunit protein eS1; v-fos transformation effector protein; v-fos transformation effector protein 1 |
| Gene Symbols | RPS3A, Rps3a1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.