Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPS3A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583853
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583853 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583853 Supplier Invitrogen™ Supplier No. PA583853
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QGTKIASDGL KGRVFEVSLA DLQNDEVAFR KFKLITEDVQ GKNCLTNFHG MDLTRDKMCS MVKKWQTMIE AHVDVKTT Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S3AE family of ribosomal proteins. It is located in the cytoplasm. Disruption of the gene encoding rat ribosomal protein S3a, also named v-fos transformation effector protein, in v-fos-transformed rat cells results in reversion of the transformed phenotype. This gene is co-transcribed with the U73A and U73B small nucleolar RNA genes, which are located in its fourth and third introns, respectively. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPS3A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RPS3A
Gene Accession No. P61247
Gene Alias 40S ribosomal protein S3a; 40S ribosomal protein S3a {ECO:0000255; 40S ribosomal protein S3b; FTE1; Fte-1; HAMAP-Rule:MF_03122}; MFTL; Protein TU-11; ribosomal protein S3a; ribosomal protein S3A1; RPS3A; Rps3a1; S3A; Small ribosomal subunit protein eS1; v-fos transformation effector protein; v-fos transformation effector protein 1
Gene Symbols RPS3A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RPS3A. Recombinant protein control fragment (Product #RP-100803).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6189
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.