Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPS4X Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP321449F
Description
RPS4X Polyclonal antibody specifically detects RPS4X in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
RPS4X | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
CCG2Single copy abundant mRNA protein, DXS306, FLJ40595, ribosomal protein S4, X-linked, RPS4cell cycle gene 2, SCAR40S ribosomal protein S4, X isoform, SCR10ribosomal protein S4X isoform, single-copy abundant mRNA | |
Produced in rabbits immunized with E. coli-derived Human RPS4X fragment | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
6191 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
FITC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction