Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPS4X Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5118843
Description
Antibody detects endogenous levels of total RPS4X.
Ribosomes, the organelles that catalyze protein synthesis, are composed of a small subunit (40S) and a large subunit (60S) that consist of over 80 distinct ribosomal proteins. Mammalian ribosomal proteins are encoded by multigene families that contain processed pseudogenes and one functional intron-containing gene within their coding regions. RPS4Y2 (ribosomal protein S4, Y-linked 2), RPS4Y1 (ribosomal protein S4, Y-linked 1) and RPS4X (ribosomal protein S4, X-linked) are proteins that contain one S4 RNA-binding domain and exist as components of the 40S ribosomal subunit, indicating a possible role in protein synthesis.
Specifications
| RPS4X | |
| Polyclonal | |
| Unconjugated | |
| RPS4X | |
| 40S ribosomal protein S4, X isoform; CCG2; cell cycle gene 2; DXS306; LOC100362640; ribosomal protein S4, X-linked; ribosomal protein S4, X-linked X-like; ribosomal protein S4X isoform; RPS4; Rps4-1; Rps4x; S4; SCAR; SCR10; single copy abundant mRNA protein; single-copy abundant mRNA; Small ribosomal subunit protein eS4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100362640, 20102, 6191 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P62701, P62702, P62703 | |
| RPS4X | |
| A synthesized peptide derived from human RPS4X(Accession P62701), corresponding to amino acid residues S91-T141. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction