Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPS4X Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575566
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Ribosomes, the organelles that catalyze protein synthesis, are composed of a small subunit (40S) and a large subunit (60S) that consist of over 80 distinct ribosomal proteins. Mammalian ribosomal proteins are encoded by multigene families that contain processed pseudogenes and one functional intron-containing gene within their coding regions. RPS4Y2 (ribosomal protein S4, Y-linked 2), RPS4Y1 (ribosomal protein S4, Y-linked 1) and RPS4X (ribosomal protein S4, X-linked) are proteins that contain one S4 RNA-binding domain and exist as components of the 40S ribosomal subunit, indicating a possible role in protein synthesis.
Specifications
RPS4X | |
Polyclonal | |
Unconjugated | |
RPS4X | |
40S ribosomal protein S4, X isoform; CCG2; cell cycle gene 2; DXS306; LOC100362640; ribosomal protein S4, X-linked; ribosomal protein S4, X-linked X-like; ribosomal protein S4X isoform; RPS4; Rps4-1; Rps4x; S4; SCAR; SCR10; single copy abundant mRNA protein; single-copy abundant mRNA; Small ribosomal subunit protein eS4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100362640, 20102, 6191 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P62701, P62702, P62703 | |
RPS4X | |
Synthetic peptide corresponding to amino acids 132-177 of Human RPS4X. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction