Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPS4X Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551468
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551468 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551468 Supplier Invitrogen™ Supplier No. PA551468
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KVRKITVGVK GIPHLVTHDA RTIRYPDPVI KVNDTVQIDL GTGKIINFIK FDTGNLCMVI GGANLGRVGV ITNRERHPGS FDVVHVKDAN GNSFATRLSN IFVIGNGNKP WISLPRGKGI RLTVA Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.

Ribosomes, the organelles that catalyze protein synthesis, are composed of a small subunit (40S) and a large subunit (60S) that consist of over 80 distinct ribosomal proteins. Mammalian ribosomal proteins are encoded by multigene families that contain processed pseudogenes and one functional intron-containing gene within their coding regions. RPS4Y2 (ribosomal protein S4, Y-linked 2), RPS4Y1 (ribosomal protein S4, Y-linked 1) and RPS4X (ribosomal protein S4, X-linked) are proteins that contain one S4 RNA-binding domain and exist as components of the 40S ribosomal subunit, indicating a possible role in protein synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPS4X
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RPS4X
Gene Accession No. P62701
Gene Alias 40S ribosomal protein S4, X isoform; CCG2; cell cycle gene 2; DXS306; LOC100362640; ribosomal protein S4, X-linked; ribosomal protein S4, X-linked X-like; ribosomal protein S4X isoform; RPS4; Rps4-1; Rps4x; S4; SCAR; SCR10; single copy abundant mRNA protein; single-copy abundant mRNA; Small ribosomal subunit protein eS4
Gene Symbols RPS4X
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RPS4Y1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6191
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.