Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPS6 Rabbit anti-Human, APC, Clone: R3A2, Abnova™

Catalog No. 89335888 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-888 10 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-888 Supplier Abnova Corporation Supplier No. MAB19054
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

RPS6 Monoclonal antibody specifically detects RPS6 in Human samples. It is validated for Flow Cyt.

Specifications

Antigen RPS6
Applications Flow Cytometry
Classification Monoclonal
Clone R3A2
Conjugate APC
Description ribosomal protein S6
Gene RPS6
Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to residues surrounding S235/S236 of human RPS6
Quantity 10 Reactions
Primary or Secondary Primary
Gene ID (Entrez) 6194
Target Species Human
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.