Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPS6KA2 Monoclonal Antibody (OTI10F11), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA809900

Description
This gene encodes a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 nonidentical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.Specifications
RPS6KA2 | |
Monoclonal | |
Unconjugated | |
RPS6KA2 | |
HU-2, MAPKAPK1C, RSK, RSK3, S6K-alpha, S6K-alpha2, p90-RSK3, p90RSK2, pp90RSK3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
6196 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
OTI10F11 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q15349 | |
RPS6KA2 | |
Human recombit protein fragment corresponding to amino acids 1-51 of human RPS6KA2 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction