Learn More
RPS6KA2 Mouse anti-Human, Clone: OTI5G2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809874

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 nonidentical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.Specifications
RPS6KA2 | |
Monoclonal | |
Unconjugated | |
RPS6KA2 | |
HU-2, MAPKAPK1C, RSK, RSK3, S6K-alpha, S6K-alpha2, p90-RSK3, p90RSK2, pp90RSK3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
6196 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI5G2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q15349 | |
RPS6KA2 | |
Human recombit protein fragment corresponding to amino acids 1-51 of human RPS6KA2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.