Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPS6KB2 Monoclonal Antibody (164CT21.2.2), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA537480

Description
RPS6KB2 (p70S6K beta) is a serine/threonine kinase with 2 non-identical kinase catalytic domains which phosphorylates the S6 ribosomal protein and eukaryotic translation initiation factor 4B (eIF4B). Phosphorylation of S6 leads to an increase in protein synthesis and cell proliferation.
Specifications
RPS6KB2 | |
Monoclonal | |
Unconjugated | |
Rps6kb2 | |
70 kDa ribosomal protein S6 kinase 2; 70kDa; KLS; p70 ribosomal S6 kinase beta; p70 S6 kinase beta; p70 S6KB; p70 S6K-beta; p70(S6K)-beta; P70-beta; P70-beta-1; P70-beta-2; p70-S6K 2; P70S6K2; p70S6Kb; ribosomal protein S6 kinase B2; ribosomal protein S6 kinase beta-2; ribosomal protein S6 kinase, 70kD, polypeptide 2; ribosomal protein S6 kinase, 70kDa, polypeptide 2; ribosomal protein S6 kinase, polypeptide 2; RPS6KB2; S6 kinase 2; S6 kinase-related kinase; S6K2; S6K-beta; S6K-beta2; S6K-beta-2; serine/threonine-protein kinase 14 beta; serine/threonine-protein kinase 14B; SRK; STK14B | |
Mouse | |
Protein G | |
RUO | |
58988, 6199 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
164CT21.2.2 | |
PBS with 0.09% sodium azide | |
Q9UBS0, Q9Z1M4 | |
Rps6kb2 | |
RPS6KB2 recombinant protein | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 κ |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction