Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPS6KB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527561
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527561 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527561 Supplier Invitrogen™ Supplier No. PA527561
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji, NIH-3T3. Predicted reactivity: Mouse (92%), Rat (92%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

RPS6KB2 (p70S6K beta) is a serine/threonine kinase with 2 non-identical kinase catalytic domains which phosphorylates the S6 ribosomal protein and eukaryotic translation initiation factor 4B (eIF4B). Phosphorylation of S6 leads to an increase in protein synthesis and cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPS6KB2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene Rps6kb2
Gene Accession No. Q9UBS0, Q9Z1M4
Gene Alias 70 kDa ribosomal protein S6 kinase 2; 70kDa; KLS; p70 ribosomal S6 kinase beta; p70 S6 kinase beta; p70 S6KB; p70 S6K-beta; p70(S6K)-beta; P70-beta; P70-beta-1; P70-beta-2; p70-S6K 2; P70S6K2; p70S6Kb; ribosomal protein S6 kinase B2; ribosomal protein S6 kinase beta-2; ribosomal protein S6 kinase, 70kD, polypeptide 2; ribosomal protein S6 kinase, 70kDa, polypeptide 2; ribosomal protein S6 kinase, polypeptide 2; RPS6KB2; S6 kinase 2; S6 kinase-related kinase; S6K2; S6K-beta; S6K-beta2; S6K-beta-2; serine/threonine-protein kinase 14 beta; serine/threonine-protein kinase 14B; SRK; STK14B
Gene Symbols Rps6kb2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human S6K2/p70 S6K beta. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58988, 6199
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.