Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPS6KC1 Monoclonal Antibody (OTI10F11)
GREENER_CHOICE

Catalog No. PIMA527060
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527060 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527060 Supplier Invitrogen™ Supplier No. MA527060
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RPS6KC1 Monoclonal Antibody for Western Blot, IHC (P)

May be involved in transmitting sphingosine-1 phosphate (SPP)-mediated signaling into the cell. Plays a role in the recruitment of PRDX3 to early endosomes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPS6KC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI10F11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RPS6KC1
Gene Accession No. Q96S38
Gene Alias 52 kDa ribosomal protein S6 kinase; AA682037; B130003F20Rik; C80612; humS6PKh1; ribosomal protein S6 kinase C1; ribosomal protein S6 kinase delta-1; ribosomal protein S6 kinase polypeptide 1; ribosomal protein S6 kinase, 52kDa, polypeptide 1; ribosomal S6 kinase-like protein with two PSK domains 118 kDa protein; RPK118; Rps6kc1; RSKL1; S6K-delta-1; S6PKh1; SPHK1-binding protein
Gene Symbols RPS6KC1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-305 of RPS6KC1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26750
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.