Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPS6KC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583197
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583197 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583197 Supplier Invitrogen™ Supplier No. PA583197
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QGLGVVESAV TANNTEESLF RICSPLSGAN EYIASTDTLK TEEVLLFTDQ TDDLAKEEPT SLFQRDSETK GESGLVLEGD KEIHQIFEDL DKKLALASRF YIPEGCIQRW A Highest antigen sequence indentity to the following orthologs: Mouse - 63%, Rat - 53%.

May be involved in transmitting sphingosine-1 phosphate (SPP)-mediated signaling into the cell. Plays a role in the recruitment of PRDX3 to early endosomes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPS6KC1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RPS6KC1
Gene Accession No. Q96S38
Gene Alias 52 kDa ribosomal protein S6 kinase; AA682037; B130003F20Rik; C80612; humS6PKh1; ribosomal protein S6 kinase C1; ribosomal protein S6 kinase delta-1; ribosomal protein S6 kinase polypeptide 1; ribosomal protein S6 kinase, 52kDa, polypeptide 1; ribosomal S6 kinase-like protein with two PSK domains 118 kDa protein; RPK118; Rps6kc1; RSKL1; S6K-delta-1; S6PKh1; SPHK1-binding protein
Gene Symbols RPS6KC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RPS6KC1. Recombinant protein control fragment (Product #RP-105858).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26750
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.