Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPS6KC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101244
Description
Antibody detects endogenous levels of total RPS6KC1.
May be involved in transmitting sphingosine-1 phosphate (SPP)-mediated signaling into the cell. Plays a role in the recruitment of PRDX3 to early endosomes.
Specifications
RPS6KC1 | |
Polyclonal | |
Unconjugated | |
RPS6KC1 | |
52 kDa ribosomal protein S6 kinase; AA682037; B130003F20Rik; C80612; humS6PKh1; ribosomal protein S6 kinase C1; ribosomal protein S6 kinase delta-1; ribosomal protein S6 kinase polypeptide 1; ribosomal protein S6 kinase, 52kDa, polypeptide 1; ribosomal S6 kinase-like protein with two PSK domains 118 kDa protein; RPK118; Rps6kc1; RSKL1; S6K-delta-1; S6PKh1; SPHK1-binding protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
26750, 320119 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8BLK9, Q96S38 | |
RPS6KC1 | |
A synthesized peptide derived from human RPS6KC1(Accession Q96S38), corresponding to amino acid residues E221-D271. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction