Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPS6KL1 Monoclonal Antibody (273CT4.5.3)
GREENER_CHOICE

Catalog No. PIMA537524
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537524 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537524 Supplier Invitrogen™ Supplier No. MA537524
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RPS6KL1 Monoclonal Antibody for Western Blot

RPS6KL1 (ribosomal protein S6 kinase-like 1) is a 549 amino acid protein that belongs to the Ser/Thr protein kinase family, S6 kinase subfamily and protein kinase superfamily. Existing as three alternatively spliced isoforms, RPS6KL1 contains one MIT domain and a protein kinase domain. The gene encoding RPS6KL1 maps to human chromosome 14, which houses over 700 genes and comprises nearly 3.5% of the human genome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPS6KL1
Applications Western Blot
Classification Monoclonal
Clone 273CT4.5.3
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene RPS6KL1
Gene Accession No. Q8R2S1, Q9Y6S9
Gene Alias A830084F09Rik; AW046440; I79_009419; ribosomal protein S6 kinase like 1; ribosomal protein S6 kinase-like 1; Rps6kl1; RSKL2
Gene Symbols RPS6KL1
Host Species Mouse
Immunogen RPS6KL1 recombinant protein.
Purification Method Euglobulin Precipitation
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100758757, 238323, 83694
Target Species Human, Mouse, Hamster
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgM κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.