Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPS7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110326
Description
Immunogen sequence: MFSSSAKIVK PNGEKPDEFE SGISQALLEL EMNSDLKAQL RELNITAAKE IEVGGGRKAI IIFVPVPQLK SFQKIQVRLV RELEKKFSGK HVVFIAQRRI LPKPTRKSRT KNKQKRPRSR TLTAVHDAIL EDLVFPSEIV GKRIRVKLDG SRLIKVHLDK AQQNNVEHKV ETFSGVYKKL TGKDVNFEFP EFQL.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S7E family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
| RPS7 | |
| Polyclonal | |
| Unconjugated | |
| RPS7 | |
| 40S ribosomal protein S7; DBA8; Mtu; ribosomal protein S7; RPS7; Rps7A; S7; S8; Small ribosomal subunit protein eS7 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20115, 29258, 6201 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.77 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P62081, P62082, P62083 | |
| RPS7 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-194 of human RPS7 (NP_001002.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction