Learn More
Invitrogen™ RPS8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531676
Description
Recommended positive controls: HepG2, 24 hpf zebrafish embryo tissue. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (93%), Chicken (97%), Sheep (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S8E family of ribosomal proteins. It is located in the cytoplasm. Increased expression of this gene in colorectal tumors and colon polyps compared to matched normal colonic mucosa has been observed. This gene is co-transcribed with the small nucleolar RNA genes U38A, U38B, U39, and U40, which are located in its fourth, fifth, first, and second introns, respectively. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
| RPS8 | |
| Polyclonal | |
| Unconjugated | |
| Rps8 | |
| 40S ribosomal protein S8; OK/SW-cl.83; ribosomal protein S8; Rps8; S8; Small ribosomal subunit protein eS8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6202 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.64 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| P62241 | |
| Rps8 | |
| Recombinant protein encompassing a sequence within the center region of human RPS8. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Zebrafish | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.