Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPS8 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA551052

Catalog No. PA551052


Only null left
Add to Cart

Description

Description

RPS8 Polyclonal Antibody detects endogenous levels of RPS8 protein.

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S8E family of ribosomal proteins. It is located in the cytoplasm. Increased expression of this gene in colorectal tumors and colon polyps compared to matched normal colonic mucosa has been observed. This gene is co-transcribed with the small nucleolar RNA genes U38A, U38B, U39, and U40, which are located in its fourth, fifth, first, and second introns, respectively. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RPS8
Polyclonal
Unconjugated
Rps8
40S ribosomal protein S8; OK/SW-cl.83; ribosomal protein S8; Rps8; S8; Small ribosomal subunit protein eS8
Rabbit
Antigen Affinity Chromatography
RUO
20116, 6202, 65136
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
P62241, P62242, P62243
Rps8
Synthesized peptide derived from the Internal region of human Ribosomal Protein S8 (aa 111-160).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.