Learn More
Invitrogen™ RPS9 Monoclonal Antibody (2D3)

Description
Peptide Sequence: VRIGVLDEGK MKLDYILGLK IEDFLERRLQ TQVFKLGLAK SIHHARVLIR QRHIRVRKQV VNIPSFIVRL DSQKHIDFSL RSPYGGGRPG.
Specifications
Specifications
| Antigen | RPS9 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 2D3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | RPS9 |
| Gene Accession No. | P46781 |
| Gene Alias | 3010033P07Rik; 40S ribosomal protein S9; AL022771; AL022885; ribosomal protein S9; Rps9; RS9; S9; Small ribosomal subunit protein uS4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.