Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPS9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561813
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561813 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561813 Supplier Invitrogen™ Supplier No. PA561813
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ELLTLDEKDP RRLFEGNALL RRLVRIGVLD EGKMKLDYIL GLKIEDFLER RLQTQVFKLG LAKSIHHARV LIRQRHIRVR KQVVNIPSFI VRLDSQKHID FSLRSPYGGG RPGRVKRKNA Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S4P family of ribosomal proteins. It is located in the cytoplasm. Variable expression of this gene in colorectal cancers compared to adjacent normal tissues has been observed, although no correlation between the level of expression and the severity of the disease has been found. As is typical for genes encoding ribosomal proteins, multiple processed pseudogenes derived from this gene are dispersed through the genome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPS9
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RPS9
Gene Accession No. P46781
Gene Alias 3010033P07Rik; 40S ribosomal protein S9; AL022771; AL022885; ribosomal protein S9; Rps9; RS9; S9; Small ribosomal subunit protein uS4
Gene Symbols RPS9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RPS9. Recombinant protein control fragment (Product #RP-102207).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6203
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.