Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPS9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104493
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104493 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104493 Supplier Thermo Scientific Supplier No. PA5104493
Add to Cart
Add to Cart

RPS9 Polyclonal Antibody

RPS9 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen RPS9
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RPS9
Gene Accession No. P29314, P46781, Q6ZWN5
Gene Alias 3010033P07Rik; 40S ribosomal protein S9; AL022771; AL022885; ribosomal protein S9; Rps9; RS9; S9; Small ribosomal subunit protein uS4
Gene Symbols RPS9
Host Species Rabbit
Immunogen A synthesized peptide derived from human RPS9(Accession P46781), corresponding to amino acid residues N39-E89.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6203, 76846, 81772
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.