Learn More
Invitrogen™ RPS9 Polyclonal Antibody

Description
RPS9 Polyclonal Antibody detects endogenous levels of RPS9 protein.
Specifications
Specifications
| Antigen | RPS9 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4 |
| Gene | RPS9 |
| Gene Accession No. | P29314, P46781, Q6ZWN5 |
| Gene Alias | 3010033P07Rik; 40S ribosomal protein S9; AL022771; AL022885; ribosomal protein S9; Rps9; RS9; S9; Small ribosomal subunit protein uS4 |
| Gene Symbols | RPS9 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.