Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPSA Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $499.50
Specifications
Antigen | RPSA |
---|---|
Concentration | 1 mg/mL |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
RPSA Polyclonal specifically detects RPSA in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
RPSA | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse | |
37 kDa laminin receptor precursor, 37/67 kDa laminin receptor, 37LRPNEM/1CHD4, 67 kDa laminin receptor, 67kD, ribosomal protein SA, 67LR, Colon carcinoma laminin-binding protein, LAMBR37 kDa laminin receptor, laminin binding protein, Laminin receptor 1, laminin receptor 1 (67kD, ribosomal protein SA), Laminin-binding protein precursor p40, lamR, LAMR 1 LAMR140S ribosomal protein SA, LBP, LBP/p40, LRP, LRP/LR, Multidrug resistance-associated protein MGr1-Ag, p40, ribosomal protein SA | |
RPSA | |
IgG | |
Affinity Purified | |
32 kDa |
1 mg/mL | |
Polyclonal | |
Rabbit | |
Core ESC Like Genes, Cytoskeleton Markers, Extracellular Matrix, Neuroscience, Stem Cell Markers | |
PBS with 0.02% Sodium Azide | |
3921 | |
Antibody was raised against a 17 amino acid peptide near the carboxy terminus of human RPSA. The immunogen is located within amino acids 190 - 240 of RPSA. Amino Acid Squence: DLYFYRDPEEIEKEEQAA | |
Primary | |
RPSA antibody is human, mouse and rat reactive. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title