Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPSA Antibody (RPSA/6332R), DyLight 594, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP314110DL594
This item is not returnable.
View return policy
Description
RPSA Monoclonal antibody specifically detects RPSA in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
RPSA | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Antibody | |
IgG κ |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
RPSA/6332R | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
37 kDa laminin receptor precursor, 37/67 kDa laminin receptor, 37LRPNEM/1CHD4, 67 kDa laminin receptor, 67kD, ribosomal protein SA, 67LR, Colon carcinoma laminin-binding protein, LAMBR37 kDa laminin receptor, laminin binding protein, Laminin receptor 1, laminin receptor 1 (67kD, ribosomal protein SA), Laminin-binding protein precursor p40, lamR, LAMR 1, LAMR140S ribosomal protein SA, LBP, LBP/p40, LRP, LRP/LR, Multidrug resistance-associated protein MGr1-Ag, p40, ribosomal protein SA | |
Recombinant full-length human RPSA protein (Uniprot: P08865) | |
0.1 mL | |
Cell Biology, Cellular Markers, Core ESC Like Genes, Extracellular Matrix, Neuroscience, Stem Cell Markers | |
3921 | |
Store at 4°C in the dark. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction