Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPSA Antibody (rRPSA/6333), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308266AF594
Description
RPSA Monoclonal antibody specifically detects RPSA in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
RPSA | |
Monoclonal | |
Alexa Fluor 594 | |
37 kDa laminin receptor precursor, 37/67 kDa laminin receptor, 37LRPNEM/1CHD4, 67 kDa laminin receptor, 67kD, ribosomal protein SA, 67LR, Colon carcinoma laminin-binding protein, LAMBR37 kDa laminin receptor, laminin binding protein, Laminin receptor 1, laminin receptor 1 (67kD, ribosomal protein SA), Laminin-binding protein precursor p40, lamR, LAMR 1, LAMR140S ribosomal protein SA, LBP, LBP/p40, LRP, LRP/LR, Multidrug resistance-associated protein MGr1-Ag, p40, ribosomal protein SA | |
Recombinant full-length human RPSA protein (Uniprot: P08865) | |
100 μg | |
Cell Biology, Cellular Markers, Core ESC Like Genes, Extracellular Matrix, Neuroscience, Stem Cell Markers | |
3921 | |
Store at 4°C in the dark. | |
IgG1 κ |
Immunohistochemistry (Paraffin) | |
rRPSA/6333 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction