Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPTOR Antibody (1B10), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP315202
This item is not returnable.
View return policy
Description
RPTOR Monoclonal antibody specifically detects RPTOR in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
RPTOR | |
Monoclonal | |
Unconjugated | |
PBS (pH 7.4), 150mM NaCl, and 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA, Immunohistochemistry | |
1B10 | |
ELISA, Immunohistochemistry 1:50-1:200 | |
KIAA1303p150 target of rapamycin (TOR)-scaffold protein containing WD-repeats, KOG1, Mip1, p150 target of rapamycin (TOR)-scaffold protein, Raptor, regulatory associated protein of MTOR, complex 1, regulatory-associated protein of mTOR | |
A synthesized peptide derived from human Raptor. UniProt ID (Q8N122). | |
100 μg | |
Autophagy, Cancer, Hypoxia, mTOR Pathway, Signal Transduction | |
57521 | |
Store at -20 to -80°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction