Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPTOR Antibody (514208) [PerCP], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB5957C
Description
RPTOR Monoclonal antibody specifically detects RPTOR in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOFSpecifications
RPTOR | |
Monoclonal | |
PerCP | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
514208 | |
Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
KIAA1303p150 target of rapamycin (TOR)-scaffold protein containing WD-repeats, KOG1, Mip1, p150 target of rapamycin (TOR)-scaffold protein, Raptor, regulatory associated protein of MTOR, complex 1, regulatory-associated protein of mTOR | |
E. coli-derived recombinant human RPTOR, Lys77-Gln230, Accession # Q8N122 | |
0.1 mL | |
Autophagy, Cancer, Hypoxia, mTOR Pathway, Signal Transduction | |
57521 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction