Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPTOR Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100766AF700
Description
RPTOR Polyclonal antibody specifically detects RPTOR in Human samples. It is validated for Western BlotSpecifications
RPTOR | |
Polyclonal | |
Western Blot | |
KIAA1303p150 target of rapamycin (TOR)-scaffold protein containing WD-repeats, KOG1, Mip1, p150 target of rapamycin (TOR)-scaffold protein, Raptor, regulatory associated protein of MTOR, complex 1, regulatory-associated protein of mTOR | |
The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human Regulatory Associated Protein of mTOR using the numbering given in entry AAM09075.1 (GeneID 57521). | |
0.1 mL | |
Autophagy, Cancer, Hypoxia, mTOR Pathway, Signal Transduction | |
57521 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction