Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPTOR Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | RPTOR |
---|---|
Dilution | Western Blot 1: 1000-1:2000, Immunohistochemistry-Paraffin 1: 100 |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224837SS
![]() |
Novus Biologicals
NBP224837SS |
0.05 mL |
Each for $181.90
|
|
|||||
NBP224837
![]() |
Novus Biologicals
NBP224837 |
0.2 mL |
Each for $487.50
|
|
|||||
Description
RPTOR Polyclonal specifically detects RPTOR in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
RPTOR | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Autophagy, Cancer, Hypoxia, mTOR Pathway, Signal Transduction | |
Whole antisera with 50 ul neat whole antisera with 0.05% Sodium Azide | |
57521 | |
A portion of amino acids 900-950 of human Raptor was used as the immunogen for this antibody. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot 1: 1000-1:2000, Immunohistochemistry-Paraffin 1: 100 | |
Polyclonal | |
Antisera | |
RUO | |
Human, Mouse | |
KIAA1303p150 target of rapamycin (TOR)-scaffold protein containing WD-repeats, KOG1, Mip1, p150 target of rapamycin (TOR)-scaffold protein, Raptor, regulatory associated protein of MTOR, complex 1, regulatory-associated protein of mTOR | |
RPTOR | |
IgG | |
Unpurified | |
The sequence used as immunogen is 86% homologous in mouse and rat. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title