Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPTOR Mouse anti-Human, Clone: 14C4, Bioss
SDP

Catalog No. 501979836
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-197-9836 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-197-9836 Supplier Bioss Supplier No. BSM51285M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Regulatory associated protein of mTOR (RAPTOR) is a binding partner of target of rapamycin (TOR). It binds to 4EBP1 and p70alpha. The binding of raptor to mTOR is essential for the mTOR-catalyzed phosphorylation of 4EBP1 in vitro. It enhances the mTOR kinase activity toward p70alpha. It is also an essential scaffold for the mTOR-catalyzed phosphorylation of 4EBP1 and mediates TOR action in vivo.

Specifications

Antigen RPTOR
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 14C4
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 40% glycerol and 0.05% sodium azide; pH
Gene RPTOR
Gene Accession No. Q8N122
Gene Alias 4932417H02Rik; KIAA1303; KOG1; Mip1; mKIAA1303; p150 target of rapamycin (TOR)-scaffold protein; p150 target of rapamycin (TOR)-scaffold protein containing WD-repeats; Rap; RAPTOR; regulatory associated protein of MTOR complex 1; regulatory associated protein of MTOR, complex 1; Regulatory-associated protein of mTOR; RGD1311784; RPTOR
Gene Symbols RPTOR
Host Species Mouse
Immunogen This RPTOR antibody is generated from a mouse immunized with recombinant protein.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57521
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.