Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPUSD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559448
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559448 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559448 Supplier Invitrogen™ Supplier No. PA559448
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VVGDLTYGEV SGREDRPFRM MLHAFYLRIP TDTECVEVCT PDPFLPSLDA CWSPHTLLQS LDQLVQALRA TPDPDPEDRG PRPGSPSALL P Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 73%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen RPUSD1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RPUSD1
Gene Accession No. Q9UJJ7
Gene Alias C16orf40; ribosomal large subunit pseudouridine synthase C-like protein; RLUCL; RNA pseudouridylate synthase domain containing 1; RNA pseudouridylate synthase domain-containing protein 1; RPUSD1
Gene Symbols RPUSD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RPUSD1. Recombinant protein control fragment (Product #RP-98612).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113000
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.