Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RPUSD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559794
Description
Immunogen sequence: GSAYRCFKER RVTKAYLALL RGHIQESRVT ISHAIGRNST EGRAHTMCIE GSQGCENPKP SLTDLVVLEH GLYAGDPVSK VLLKPLTG Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 86%.

Specifications
RPUSD1 | |
Polyclonal | |
Unconjugated | |
RPUSD1 | |
C16orf40; ribosomal large subunit pseudouridine synthase C-like protein; RLUCL; RNA pseudouridylate synthase domain containing 1; RNA pseudouridylate synthase domain-containing protein 1; RPUSD1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
113000 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UJJ7 | |
RPUSD1 | |
Recombinant protein corresponding to Human RPUSD1. Recombinant protein control fragment (Product #RP-98613). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction