Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPUSD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558000
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558000 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558000 Supplier Invitrogen™ Supplier No. PA558000
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VHMVLQLCPV LGDHMYSARV GTVLGQRFLL PAENNKPQRQ VLDEALLRRL HLTPSQAAQL PLHLHLHRLL LPGTR Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 84%.

RNA Pseudouridine Synthase D3 (RPUSD3) is a gene that encodes a pseudouridine synthase, an enzyme responsible for catalyzing the isomerization of uridine to pseudouridine in RNA molecules. This post-transcriptional modification is crucial for the stability and function of tRNAs and rRNAs. RPUSD3, along with other pseudouridine synthases such as PUS1, PUSL1, and TRUB2, is predicted to localize to mitochondria, indicating its role in mitochondrial RNA processing and maturation. The pseudouridylation activity provided by RPUSD3 is essential for the proper folding and functionality of mitochondrial RNAs, which are fundamental for the biogenesis of the mitochondrial electron transport chain and ATP synthase. While detailed structural insights into RPUSD3 are not yet fully characterized, its enzymatic function is critical in maintaining cellular energy metabolism and overall mitochondrial health.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPUSD3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RPUSD3
Gene Accession No. Q6P087
Gene Alias AI527266; Mitochondrial mRNA pseudouridine synthase RPUSD3; RGD1307582; RNA pseudouridylate synthase domain containing 3; RNA pseudouridylate synthase domain-containing protein 3; Rpusd3
Gene Symbols RPUSD3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RPUSD3. Recombinant protein control fragment (Product #RP-96383).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285367
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.