Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RPUSD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558885
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558885 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558885 Supplier Invitrogen™ Supplier No. PA558885
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NLVVINKPYG LPVHGGPGVQ LCITDVLPIL AKMLHGHKAE PLHLCHRLDK ETTGVMVLAW DKDMAHQVQE LFRTRQVVKK YWAITVHVP Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen RPUSD4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RPUSD4
Gene Accession No. Q96CM3
Gene Alias 2410001E19Rik; Mitochondrial RNA pseudouridine synthase Rpusd4; RGD1311268; RNA pseudouridylate synthase domain containing 4; RNA pseudouridylate synthase domain-containing protein 4; Rpusd4
Gene Symbols RPUSD4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RPUSD4. Recombinant protein control fragment (Product #RP-97431).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84881
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.