Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ RQ 00203078

Catalog No. 538810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-881-0 10 mg
53-885-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 53-881-0 Supplier Tocris Bioscience™ Supplier No. 5388/10
This item is not returnable. View return policy

Potent and selective TRPM8 antagonist; orally active

RQ 00203078 is a potent TRPM8 antagonist (IC50 values are 5.3 and 8.3 nM for rat and human channels respectively). Exhibits >350-fold selectivity for TRPM8 over TRPV4, TRPV1 and TRPA1. Reduces HSC3 and HSC4 oral squamous carcinoma cell migration and invasion in vitro. Also attenuates icilin -induced wet dog shakes in rats. Orally active.

Specifications

Quantity 10 mg
Product Type RQ 00203078
Purity >98%
Molecular Weight (g/mol) 554.85

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.